3-Fluoro-2-methoxybenzoic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-3321-1G
HC-3321-5G
COBBHC-3321-1GEA
0
GBP
COBBHC-3321-1G
COBBHC-3321-5G
3-Fluoro-2-methoxybenzoic acid
3-Fluoro-2-methoxybenzoic acid
Formula:
C₈H₇FO₃ MW: 170.14 g/mol |
MDL Number:
MFCD00973972 CAS Number: 106428-05-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...