3-Fluoro-o-anisaldehyde

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-3173-5G HC-3173-1G
COBBHC-3173-5GEA 0 GBP
COBBHC-3173-5G COBBHC-3173-1G
3-Fluoro-o-anisaldehyde
3-Fluoro-o-anisaldehyde
Formula: C₈H₇FO₂
MDL Number: MFCD04112486
CAS Number: 74266-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR