3-Ethoxy-2-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8018-1G BB-8018-5G
COBBBB-8018-1GEA 0 GBP
COBBBB-8018-1G COBBBB-8018-5G
3-Ethoxy-2-fluorophenylboronic acid
3-Ethoxy-2-fluorophenylboronic acid
Formula: C₈H₁₀BFO₃
MDL Number: MFCD06798077
CAS Number: 855230-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR