3-Cyanophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (3-Cyanophenyl)metaboric acid, Dihydroxy(3-cyanophenyl)borane, 3-Cyanobenzeneboronic acid

BB-2455-25G BB-2455-1G BB-2455-100G BB-2455-5G
COBBBB-2455-25GEA 0 GBP
COBBBB-2455-25G COBBBB-2455-1G COBBBB-2455-100G COBBBB-2455-5G
3-Cyanophenylboronic acid
3-Cyanophenylboronic acid
Formula: C₇H₆BNO₂
MW: 146.94 g/mol
MDL Number: MFCD01318967
CAS Number: 150255-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR