3-Chlorothiobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021322-1G 021322-25G 021322-5G
FLUO021322-1GEA 0 GBP
FLUO021322-1G FLUO021322-25G FLUO021322-5G
3-Chlorothiobenzamide
3-Chlorothiobenzamide
Formula: C₇H₆ClNS
MDL Number: MFCD00276993
CAS Number: 2548-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR