5-Chloro-2-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Chloro-2-fluorobenzeneboronic acid, (5-Chloro-2-fluorophenyl)metaboric acid, (5-Chloro-2-fluorophenyl)boronic acid, Dihydroxy(5-chloro-2-fluorophenyl)borane

BB-2388-5G BB-2388-25G BB-2388-1G
COBBBB-2388-5GEA 0 GBP
COBBBB-2388-5G COBBBB-2388-25G COBBBB-2388-1G
5-Chloro-2-fluorophenylboronic acid
5-Chloro-2-fluorophenylboronic acid
Formula: C₆H₅BClFO₂
MDL Number: MFCD05664225
CAS Number: 352535-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR