5-Chloro-2-fluorobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CA-4013-5G CA-4013-25G CA-4013-100G
COBBCA-4013-5GEA 0 GBP
COBBCA-4013-5G COBBCA-4013-25G COBBCA-4013-100G
5-Chloro-2-fluorobenzoic acid
5-Chloro-2-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MDL Number: MFCD00665762
CAS Number: 394-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR