3-Chloro-5-nitrobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CA-4197-5G CA-4197-1G
COBBCA-4197-5GEA 0 GBP
COBBCA-4197-5G COBBCA-4197-1G
3-Chloro-5-nitrobenzoic acid
3-Chloro-5-nitrobenzoic acid
Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
MDL Number: MFCD09263224
CAS Number: 34662-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR