3-Chloro-4-nitrobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CA-4455-5G CA-4455-1G CA-4455-25G
COBBCA-4455-5GEA 0 GBP
COBBCA-4455-5G COBBCA-4455-1G COBBCA-4455-25G
3-Chloro-4-nitrobenzoic acid
3-Chloro-4-nitrobenzoic acid
Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
MDL Number: MFCD01632172
CAS Number: 39608-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR