3-Chloro-4-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Chloro-4-fluorobenzeneboronic acid, (3-Chloro-4-fluorophenyl)metaboric acid, Dihydroxy(3-chloro-4-fluorophenyl)borane

BB-2656-5G BB-2656-25G BB-2656-100G
COBBBB-2656-5GEA 0 GBP
COBBBB-2656-5G COBBBB-2656-25G COBBBB-2656-100G
3-Chloro-4-fluorophenylboronic acid
3-Chloro-4-fluorophenylboronic acid
Formula: C₆H₅BClFO₂
MDL Number: MFCD00051800
CAS Number: 144432-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR