3-Chloro-2-fluorobenzenesulphonamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021394-5G 021394-1G
FLUO021394-5GEA 0 GBP
FLUO021394-5G FLUO021394-1G
3-Chloro-2-fluorobenzenesulphonamide
3-Chloro-2-fluorobenzenesulphonamide
Formula: C₆H₅ClFNO₂S
MW: 209.63 g/mol
CAS Number: 351003-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR