3-Bromothiobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021312-1G 021312-5G
FLUO021312-1GEA 0 GBP
FLUO021312-1G FLUO021312-5G
3-Bromothiobenzamide
3-Bromothiobenzamide
Formula: C₇H₆BrNS
MDL Number: MFCD04973313
CAS Number: 2227-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR