3-Bromofuran

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0730-5G OT-0730-25G
COBBOT-0730-5GEA 0 GBP
COBBOT-0730-5G COBBOT-0730-25G
3-Bromofuran
3-Bromofuran
Formula: C₄H₃BrO
Boiling Pt: 103 °C (1013 hPa)
Density: 1,6957 g/cm³ (0 °C)
MDL Number: MFCD00005347
CAS Number: 22037-28-1
EINECS: 244-744-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR