3-Bromoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PY-7143-100G PY-7143-25G PY-7143-5G
COBBPY-7143-100GEA 0 GBP
COBBPY-7143-100G COBBPY-7143-25G COBBPY-7143-5G
3-Bromoquinoline
3-Bromoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 274 °C (1013 hPa)
Melting Pt: 15 °C
Density: 1,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006767
CAS Number: 5332-24-1
EINECS: 226-237-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR