3-Bromobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CA-4012-5G CA-4012-25G CA-4012-100G
COBBCA-4012-5GEA 0 GBP
COBBCA-4012-5G COBBCA-4012-25G COBBCA-4012-100G
3-Bromobenzoic acid
3-Bromobenzoic acid
Formula: C₇H₅BrO₂
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 155 °C
Density: 1,845 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002487
CAS Number: 585-76-2
EINECS: 209-562-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR