5-Bromo-2-fluorobenzaldehyde
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OS-7553-25G
OS-7553-100G
COBBOS-7553-25GEA
0
GBP
COBBOS-7553-25G
COBBOS-7553-100G
5-Bromo-2-fluorobenzaldehyde
5-Bromo-2-fluorobenzaldehyde
Formula:
C₇H₄BrFO MW: 203.01 g/mol Boiling Pt: 229 Melting Pt: 49 Density: 1.71 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00070755 CAS Number: 93777-26-5 EINECS: 298-056-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...