3-Bromo-6-chloropyridazine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0551-5G
OT-0551-1G
COBBOT-0551-5GEA
0
GBP
COBBOT-0551-5G
COBBOT-0551-1G
3-Bromo-6-chloropyridazine
3-Bromo-6-chloropyridazine
Formula:
C₄H₂BrClN₂ |
MDL Number:
MFCD09834814 CAS Number: 89089-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...