3-Bromo-5-fluorobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4113-5G CA-4113-25G CA-4113-100G
COBBCA-4113-5GEA 0 GBP
COBBCA-4113-5G COBBCA-4113-25G COBBCA-4113-100G
3-Bromo-5-fluorobenzoic acid
3-Bromo-5-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD01569540
CAS Number: 176548-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR