5-Bromo-2,3-dichloropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7870-25G
PY-7870-5G
PY-7870-100G
COBBPY-7870-25GEA
0
GBP
COBBPY-7870-25G
COBBPY-7870-5G
COBBPY-7870-100G
5-Bromo-2,3-dichloropyridine
5-Bromo-2,3-dichloropyridine
Formula:
C₅H₂BrCl₂N MW: 226.89 g/mol |
MDL Number:
MFCD04112492 CAS Number: 97966-00-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...