3-Bromo-5-(trifluoromethyl)benzaldehyde

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromo-5-trifluoromethylbenzaldehyde

OT-0424-5G OT-0424-25G OT-0424-1G
COBBOT-0424-5GEA 0 GBP
COBBOT-0424-5G COBBOT-0424-25G COBBOT-0424-1G
3-Bromo-5-(trifluoromethyl)benzaldehyde
3-Bromo-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₄BrF₃O
MDL Number: MFCD08059506
CAS Number: 477535-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR