3-Bromo-4-fluorobenzoic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

COBBCA-4004-25GEA 0 GBP
COBBCA-4004-25G COBBCA-4004-100G COBBCA-4004-5G
3-Bromo-4-fluorobenzoic acid
3-Bromo-4-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD00042463
CAS Number: 1007-16-5
EINECS: 213-751-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR