3-Bromo-o-toluic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4591-100G CA-4591-25G CA-4591-5G
COBBCA-4591-100GEA 0 GBP
COBBCA-4591-100G COBBCA-4591-25G COBBCA-4591-5G
3-Bromo-o-toluic acid
3-Bromo-o-toluic acid
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
MDL Number: MFCD00270097
CAS Number: 76006-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR