3-Bromo-2-methoxy-5-nitropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7712-100G PY-7712-1G PY-7712-25G PY-7712-5G
COBBPY-7712-100GEA 0 GBP
COBBPY-7712-100G COBBPY-7712-1G COBBPY-7712-25G COBBPY-7712-5G
3-Bromo-2-methoxy-5-nitropyridine
3-Bromo-2-methoxy-5-nitropyridine
Formula: C₆H₅BrN₂O₃
MDL Number: MFCD06659517
CAS Number: 15862-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR