3-Bromo-2-fluoropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7408-25G OS-7408-5G OS-7408-1G
COBBOS-7408-25GEA 0 GBP
COBBOS-7408-25G COBBOS-7408-5G COBBOS-7408-1G
3-Bromo-2-fluoropyridine
3-Bromo-2-fluoropyridine
Formula: C₅H₃BrFN
MDL Number: MFCD04112496
CAS Number: 36178-05-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR