3-Aminothiobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Aminobenzothioamide

021308-1G 021308-2.5G
FLUO021308-1GEA 0 GBP
FLUO021308-1G FLUO021308-2.5G
3-Aminothiobenzamide
3-Aminothiobenzamide
Formula: C₇H₈N₂S
MDL Number: MFCD04973325
CAS Number: 78950-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR