3-Amino-o-toluic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Amino-2-methylbenzoic acid

CA-4469-25G CA-4469-100G
COBBCA-4469-25GEA 0 GBP
COBBCA-4469-25G COBBCA-4469-100G
3-Amino-o-toluic acid
3-Amino-o-toluic acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 347
Melting Pt: 181
Storage Temperature: Ambient
MDL Number: MFCD00075026
CAS Number: 52130-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR