3-Amino-2-chloropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-3-pyridylamine, 2-Chloropyridin-3-amine

BK-4502-25G BK-4502-100G
COBBBK-4502-25GEA 0 GBP
COBBBK-4502-25G COBBBK-4502-100G
3-Amino-2-chloropyridine
3-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 78…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006238
CAS Number: 6298-19-7
EINECS: 228-572-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR