3-Amino-2,5-dichloropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7918-5G
PY-7918-1G
PY-7918-25G
COBBPY-7918-5GEA
0
GBP
COBBPY-7918-5G
COBBPY-7918-1G
COBBPY-7918-25G
3-Amino-2,5-dichloropyridine
3-Amino-2,5-dichloropyridine
Formula:
C₅H₄Cl₂N₂ MW: 163.01 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00204157 CAS Number: 78607-32-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...