3,5-Difluorobenzoic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
CA-4029-25G
CA-4029-100G
COBBCA-4029-25GEA
0
GBP
COBBCA-4029-25G
COBBCA-4029-100G
3,5-Difluorobenzoic acid
3,5-Difluorobenzoic acid
Formula:
C₇H₄F₂O₂ Boiling Pt: 242 °C (1013 hPa) Melting Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010323 CAS Number: 455-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...