3,5-Dibromophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

BB-2661-25G BB-2661-1G BB-2661-5G
COBBBB-2661-25GEA 0 GBP
COBBBB-2661-25G COBBBB-2661-1G COBBBB-2661-5G
3,5-Dibromophenylboronic acid
3,5-Dibromophenylboronic acid
Formula: C₆H₅BBr₂O₂
MW: 279.72 g/mol
MDL Number: MFCD01075725
CAS Number: 117695-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR