Acetonitrile
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
8825.1
8825.2
ROTH8825.1EA
0
GBP
ROTH8825.1
ROTH8825.2
Acetonitrile
Acetonitrile
Formula:
H₃CCN Boiling Pt: 81.6 °C (1013 hPa) Melting Pt: –45.7 °C Density: 0.782 g/cm³ (20 °C) Flash Pt: 2 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...