3,4-Dichloropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7597-5G
OS-7597-25G
OS-7597-1G
COBBOS-7597-5GEA
0
GBP
COBBOS-7597-5G
COBBOS-7597-25G
COBBOS-7597-1G
3,4-Dichloropyridine
3,4-Dichloropyridine
Formula:
C₅H₃Cl₂N |
MDL Number:
MFCD01861989 CAS Number: 55934-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...