3,4,5-Trimethoxybenzamide ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

L12155.14 L12155.22
L12155.14EA 29.5 GBP
L12155.14 L12155.22
3,4,5-Trimethoxybenzamide ≥98%
3,4,5-Trimethoxybenzamide

Purity: 98% CAS No.: 3086-62-2 Molecular Formula: C10H13NO4 Formula Weight: 211.22 Melting Point: 178-180° Beilstein Reference: 2697325

Formula: C₁₀H₁₃NO₄
Melting Pt: 178…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00017129
CAS Number: 3086-62-2
EINECS: 221-406-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR