3,3,4,5,5,5-Hexafluoro-2-pentanol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010478-10G 010478-1G 010478-5G
FLUO010478-10GEA 0 GBP
FLUO010478-10G FLUO010478-1G FLUO010478-5G
3,3,4,5,5,5-Hexafluoro-2-pentanol
3,3,4,5,5,5-Hexafluoro-2-pentanol
Formula: C₅H₆F₆O
MDL Number: MFCD00155852
CAS Number: 2711-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR