3-(Trifluoromethyl)-1H-pyrazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF183.10 CFF183.25 CFF183.50 007661-5G 007661-1G
CHMPCFF183.10EA 0 GBP
CHMPCFF183.10 CHMPCFF183.25 CHMPCFF183.50 FLUO007661-5G FLUO007661-1G
3-(Trifluoromethyl)-1H-pyrazole
3-(Trifluoromethyl)-1H-pyrazole
Formula: C₄H₃F₃N₂
MDL Number: MFCD00115018
CAS Number: 20154-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR