3-(Trifluoromethoxy)benzoic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OS-7503-5G
OS-7503-25G
OS-7503-100G
COBBOS-7503-5GEA
0
GBP
COBBOS-7503-5G
COBBOS-7503-25G
COBBOS-7503-100G
3-(Trifluoromethoxy)benzoic acid
3-(Trifluoromethoxy)benzoic acid
Formula:
C₈H₅F₃O₃ Boiling Pt: 272 °C (1013 hPa) Melting Pt: 91…92 °C |
MDL Number:
MFCD00041501 CAS Number: 1014-81-9 EINECS: 213-802-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...