3-(3-Bromophenyl)propionic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4673-25G
CA-4673-1G
CA-4673-5G
COBBCA-4673-25GEA
0
GBP
COBBCA-4673-25G
COBBCA-4673-1G
COBBCA-4673-5G
3-(3-Bromophenyl)propionic acid
3-(3-Bromophenyl)propionic acid
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol |
MDL Number:
MFCD01310792 CAS Number: 42287-90-1 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...