2-Methoxy-5-nitropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PY-7303-100G
PY-7303-25G
COBBPY-7303-100GEA
0
GBP
COBBPY-7303-100G
COBBPY-7303-25G
2-Methoxy-5-nitropyridine
2-Methoxy-5-nitropyridine
Formula:
C₆H₆N₂O₃ Melting Pt: 114 °C |
MDL Number:
MFCD00006263 CAS Number: 5446-92-4 EINECS: 226-661-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...