2-Iodo-m-toluic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2844-5G HC-2844-25G HC-2844-1G
COBBHC-2844-5GEA 0 GBP
COBBHC-2844-5G COBBHC-2844-25G COBBHC-2844-1G
2-Iodo-m-toluic acid
2-Iodo-m-toluic acid
Formula: C₈H₇IO₂
MW: 262.05 g/mol
MDL Number: MFCD00079764
CAS Number: 108078-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR