2-Hydroxybenzimidazole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002990-5G
002990-25G
002990-10G
FLUO002990-5GEA
0
GBP
FLUO002990-5G
FLUO002990-25G
FLUO002990-10G
2-Hydroxybenzimidazole
2-Hydroxybenzimidazole
Formula:
C₇H₆N₂O Melting Pt: 320…322 °C Density: 1,42 g/cm³ (20 °C) |
MDL Number:
MFCD00127894 CAS Number: 615-16-7 EINECS: 210-412-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...