2-Fluoro-6-(trifluoromethyl)pyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4316-100G
CA-4316-5G
CA-4316-25G
CA-4316-1G
COBBCA-4316-100GEA
0
GBP
COBBCA-4316-100G
COBBCA-4316-5G
COBBCA-4316-25G
COBBCA-4316-1G
2-Fluoro-6-(trifluoromethyl)pyridine
2-Fluoro-6-(trifluoromethyl)pyridine
Formula:
C₆H₃F₄N MW: 165.09 g/mol |
MDL Number:
MFCD03092901 CAS Number: 94239-04-0 EINECS: 428-100-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...