2-Fluoro-5-nitropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7446-5G
OS-7446-25G
OS-7446-1G
COBBOS-7446-5GEA
0
GBP
COBBOS-7446-5G
COBBOS-7446-25G
COBBOS-7446-1G
2-Fluoro-5-nitropyridine
2-Fluoro-5-nitropyridine
Formula:
C₅H₃FN₂O₂ |
MDL Number:
MFCD03095059 CAS Number: 456-24-6 EINECS: 207-260-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...