2-Fluoro-5-nitrobenzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009221-5G
009221-25G
FLUO009221-5GEA
0
GBP
FLUO009221-5G
FLUO009221-25G
2-Fluoro-5-nitrobenzaldehyde
2-Fluoro-5-nitrobenzaldehyde
Formula:
C₇H₄FNO₃ MW: 169.11 g/mol |
MDL Number:
MFCD00042298 CAS Number: 27996-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...