2-Fluoro-4-methylbenzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-4-methylbenzene-1-sulphonyl chloride

021396-1G 021396-5G 021396-25G
FLUO021396-1GEA 0 GBP
FLUO021396-1G FLUO021396-5G FLUO021396-25G
2-Fluoro-4-methylbenzenesulphonyl chloride
2-Fluoro-4-methylbenzenesulphonyl chloride
Formula: C₇H₆ClFO₂S
MW: 208.64 g/mol
MDL Number: MFCD03094330
CAS Number: 518070-29-6
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR