2-Fluoro-4-(trifluoromethyl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1455-1G PY-1455-25G PY-1455-5G
COBBPY-1455-1GEA 0 GBP
COBBPY-1455-1G COBBPY-1455-25G COBBPY-1455-5G
2-Fluoro-4-(trifluoromethyl)pyridine
2-Fluoro-4-(trifluoromethyl)pyridine
Formula: C₆H₃F₄N
MDL Number: MFCD09879262
CAS Number: 118078-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR