1,1,1-Tris(hydroxymethyl)propane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1,1,1-Trimethylolpropane, Propylidynetrimethanol, Trimethylol propane, Propylidynetris(methanol), 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol
93370-1KG
93370-250G
SIAL93370-1KGEA
107
GBP
SIAL93370-1KG
SIAL93370-250G
1,1,1-Tris(hydroxymethyl)propane, Sigma-Aldrich®
1,1,1-Tris(hydroxymethyl)propane
Formula:
C₆H₁₄O₃ MW: 134.18 g/mol Boiling Pt: 296 Melting Pt: 53 Density: 1.15 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004694 CAS Number: 77-99-6 EINECS: 201-074-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...