2-Ethoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2597-25G BB-2597-1G BB-2597-5G
COBBBB-2597-25GEA 0 GBP
COBBBB-2597-25G COBBBB-2597-1G COBBBB-2597-5G
2-Ethoxyphenylboronic acid
2-Ethoxyphenylboronic acid
Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
MDL Number: MFCD00674027
CAS Number: 213211-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR