2-Vinylpyridine ≥97% stabilised
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Stabilisation:
Stabilised with 4-tert-butylpyrocatechol (TBC)
A14056.AE
A14056.AP
A14056.AEEA
32.4
GBP
A14056.AE
A14056.AP
2-Vinylpyridine ≥97% stabilised
2-Vinylpyridine
Stabilized with 0.1% 4-tert-butylcatechol
Purity: 97%
CAS No.: 100-69-6
Molecular Formula: C7H7N
Formula Weight: 105.14
Storage Temperature: 2°C to 8°C
Flash Point: 47°(117°F)
Refractive Index: 1.5500
Odour: Pungent
Melting Point: -50°
Boiling Point: 159°
Beilstein Reference: 104505
UN No.: 3073
MDL No.: MFCD00006355
Formula:
C₇H₇N MW: 105.14 g/mol Boiling Pt: 159 °C Melting Pt: –50 °C Density: 0.974 Flash Pt: 47 °C (117 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006355 CAS Number: 100-69-6 EINECS: 202-879-8 UN: 3073 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...