2-Thiophenecarbonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Thienyl cyanide, Thiophene-2-carbonitrile, 2-Cyanothiophene
OS-7671-25G
OS-7671-100G
COBBOS-7671-25GEA
0
GBP
COBBOS-7671-25G
COBBOS-7671-100G
2-Thiophenecarbonitrile
2-Thiophenecarbonitrile
Formula:
C₅H₃NS MW: 109.15 g/mol Boiling Pt: 190…192 °C (1013 hPa) Density: 1.18 g/cm³ (15 °C) |
MDL Number:
MFCD00005416 CAS Number: 1003-31-2 EINECS: 213-706-0 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...