2-Cyanophenylboronic acid 96%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
(2-Cyanophenyl)metaboric acid, 2-Cyanobenzeneboronic acid, (2-Cyanophenyl)boronic acid, Dihydroxy(2-cyanophenyl)borane
H52609.03
H52609.06
H52609.03EA
49.6
GBP
H52609.03
H52609.06
2-Cyanophenylboronic acid 96%
2-Cyanophenylboronic acid
Purity: 96%
CAS No.: 138642-62-3
Molecular Formula: C7H6BNO2
Formula Weight: 146.94
Melting Point: 250-254°
MDL No.: MFCD01632208
Formula:
C₇H₆BNO₂ MW: 146.94 g/mol Melting Pt: 250…254 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632208 CAS Number: 138642-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...